Learn More
STEMCELL Technologies UM729, Size: 250 µg
SDP
Catalog No. NC2075739
Encompass
UM729 is a pyrimido-[4,5-b]-indole derivative which enhances the self-renewal of human hematopoietic stem cells in vitro. UM729 does not inhibit the aryl hydrocarbon receptor (AHR) pathway, but has been shown to collaborate with AHR antagonists in preventing differentiation of acute myeloid leukemia (AML) cells in culture.MAINTENANCE AND SELF-RENEWAL· Enhances human hematopoietic stem cell self-renewal in vitro (Fares et al.).CANCER RESEARCH:· Collaborates with StemRegenin 1 (SR1) in preventing differentiation of AML cells in culture (Pabst et al.).For clinical use of UM729, please contact ExCellThera.
Catalog No. NC2075739 Supplier STEMCELL Technologies Supplier No. 72332
UM729 is a pyrimido-[4,5-b]-indole derivative which enhances the self-renewal of human hematopoietic stem cells in vitro. UM729 does not inhibit the aryl hydrocarbon receptor (AHR) pathway, but has been shown to collaborate with AHR antagonists in preventing differentiation of acute myeloid leukemia (AML) cells in culture.MAINTENANCE AND SELF-RENEWAL· Enhances human hematopoietic stem cell self-renewal in vitro (Fares et al.).CANCER RESEARCH:· Collaborates with StemRegenin 1 (SR1) in preventing differentiation of AML cells in culture (Pabst et al.).For clinical use of UM729, please contact ExCellThera.