Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Umbelliferone, Analytical Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015926 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5926 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5926 Supplier MilliporeSigma Supelco Supplier No. 5482650MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Umbelliferone is a naturally occurring coumarin, present in a variety of medicinal plants from the Apiaceae (Umbelliferae) family and also in plants from other families such as mouse-ear hawkweed.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 93-35-6
Physical Form Neat
Linear Formula C9H6O3
MDL Number MFCD00006878
Melting Point 230°C (decomposition)
Synonym 7-Hydroxy-2 H-1-benzopyran-2-one; 7-Hydroxycoumarin
Quantity 50 mg
Formula Weight 162.14
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.