Learn More
AdipoGen Umbellulone
SDP
Supplier:  AdipoGen AGCN20085M100
Encompass_Preferred
Chemical. CAS 546-78-1. Formula C10H14O. MW 150.2. Isolated from Umbellularia californica Nutt. Offensive principle of the so-called "headache tree". Causes a painful cold sensation. Selective TRPA1 activator. Weak TRPM8 activator. Releases CGRP calcitonin gene-related peptide after TRPA1 activation which leads to activation of trigeminovascular pathways and CGRP-mediated neurogenic vasodilatation relevant to produce headache/migraine.
Catalog No. 50-256-2667