Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Umbralisib | 1532533-67-7 | >98.0% | 25 MG
SDP

Catalog No. 5000230156
Encompass_Preferred

Umbralisib is a small-molecule dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor used as a research reagent for preclinical studies in oncology and cell signaling.

  • Dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibition for pathway and target validation.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO at 25 mg/mL (ultrasonic) for preparation of stock solutions.
  • Molecular weight 571.55 g/mol and formula C31H24F3N5O3.
  • Intended for research use in preclinical or laboratory applications.

Catalog No. 50-002-30156 Supplier Medchemexpress LLC Supplier No. HY1227925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Umbralisib is a small-molecule dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor used as a research reagent for preclinical studies in oncology and cell signaling.

  • Dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibition for pathway and target validation.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO at 25 mg/mL (ultrasonic) for preparation of stock solutions.
  • Molecular weight 571.55 g/mol and formula C31H24F3N5O3.
  • Intended for research use in preclinical or laboratory applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.