Learn More
Medchemexpress LLC Umbralisib | 1532533-67-7 | 98.9% | 50 MG
SDP
Catalog No. 5000230410
Encompass_Preferred
Umbralisib is an orally active, selective inhibitor of PI3Kδ and casein kinase-1-ε used in preclinical research on B-cell malignancies and kinase signaling. Supplied as a white to off-white solid with high purity for laboratory assays.
  • Selective PI3Kδ and CK1ε inhibition for oncology research.
  • High purity (≈98.9%) solid suitable for in vitro and preclinical studies.
  • Recommended storage: powder stable at -20°C for long-term storage.
  • Supplied in a 50 mg package for assay development and compound screening.
Catalog No. 50-002-30410 Supplier Medchemexpress LLC Supplier No. HY1227950MG
Umbralisib is an orally active, selective inhibitor of PI3Kδ and casein kinase-1-ε used in preclinical research on B-cell malignancies and kinase signaling. Supplied as a white to off-white solid with high purity for laboratory assays.
  • Selective PI3Kδ and CK1ε inhibition for oncology research.
  • High purity (≈98.9%) solid suitable for in vitro and preclinical studies.
  • Recommended storage: powder stable at -20°C for long-term storage.
  • Supplied in a 50 mg package for assay development and compound screening.