Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Umbralisib R-enantiomer | 1532533-69-9 | 99.5% | 5 MG
SDP

Catalog No. 5000231601
Encompass_Preferred

Umbralisib R-enantiomer is the R-enantiomer of umbralisib, a PI3Kδ inhibitor supplied for research use. It is reported as the less-active enantiomer and is intended for biochemical and preclinical studies.

  • Acts as a PI3Kδ inhibitor for research applications.
  • High purity suitable for analytical and preclinical work.
  • Molecular weight 571.55 g/mol and formula C31H24F3N5O3.
  • Appearance solid, white to light yellow.
  • Available in small research quantities, including 5 mg packages.
  • Provided with CAS number for unambiguous identification.

Catalog No. 50-002-31601 Supplier Medchemexpress LLC Supplier No. HY12279F5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Umbralisib R-enantiomer is the R-enantiomer of umbralisib, a PI3Kδ inhibitor supplied for research use. It is reported as the less-active enantiomer and is intended for biochemical and preclinical studies.

  • Acts as a PI3Kδ inhibitor for research applications.
  • High purity suitable for analytical and preclinical work.
  • Molecular weight 571.55 g/mol and formula C31H24F3N5O3.
  • Appearance solid, white to light yellow.
  • Available in small research quantities, including 5 mg packages.
  • Provided with CAS number for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.