Learn More
TARGETMOL CHEMICALS INC UMBRALISIB R-ENANTIOMER 5MG
SDP
Catalog No. 502581376
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Umbralisib R-enantiomer (RP5264 R-enantiomer) is a delta inhibitor of PI3K and a less active enantiomer of TGR-1202. The IC50 of Umbralisib R-enantiomers against (delta)PI3 kinase is at least 20 times lower than that of the inhibitors against other PI3K isoforms (A (beta) and (gamma)). purity: 95%
Catalog No. 50-258-1376 Supplier TARGETMOL CHEMICALS INC Supplier No. T131405MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Umbralisib R-enantiomer (RP5264 R-enantiomer) is a delta inhibitor of PI3K and a less active enantiomer of TGR-1202. The IC50 of Umbralisib R-enantiomers against (delta)PI3 kinase is at least 20 times lower than that of the inhibitors against other PI3K isoforms (A (beta) and (gamma)). purity: 95%