Learn More
Selleck Chemical LLC Umeclidinium bromide S3735-5mg
SDP
Catalog No. 503109174
Encompass_Preferred
Umeclidinium bromide(GSK573719A) is a long-acting muscarinic antagonist approved for the maintenance treatment of chronic obstructive pulmonary disease (COPD)
Catalog No. 50-310-9174 Supplier Selleck Chemical LLC Supplier No. S37355MG
Umeclidinium bromide(GSK573719A) is a long-acting muscarinic antagonist approved for the maintenance treatment of chronic obstructive pulmonary disease (COPD)