Learn More
Medchemexpress LLC Umeclidinium bromide | 869113-09-7 | 508.49 | 100 MG
SDP
Catalog No. 5000449230
Encompass_Preferred
Umeclidinium bromide is a novel mAChR antagonist with an affinity (Ki) for cloned human M1-M5 mAChRs ranging from 0.05 to 0.16 nM. It inhibits the human ether-a-go-go-related gene channel tail current in a concentration-dependent manner.
  • Novel mAChR antagonist
  • Inhibits human ether-a-go-go-related gene channel tail current
  • Shows slow functional reversibility at cloned human M3 mAChRs
  • Shows slow functional reversibility at endogenous mAChR in isolated human bronchus
  • For research use only
Catalog No. 50-004-49230 Supplier Medchemexpress LLC Supplier No. HYW004415100MG
Umeclidinium bromide is a novel mAChR antagonist with an affinity (Ki) for cloned human M1-M5 mAChRs ranging from 0.05 to 0.16 nM. It inhibits the human ether-a-go-go-related gene channel tail current in a concentration-dependent manner.
  • Novel mAChR antagonist
  • Inhibits human ether-a-go-go-related gene channel tail current
  • Shows slow functional reversibility at cloned human M3 mAChRs
  • Shows slow functional reversibility at endogenous mAChR in isolated human bronchus
  • For research use only