Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Umeclidinium bromide | 869113-09-7 | 99.1% | 1 ML
SDP

Catalog No. 5000232730
Encompass_Preferred

Umeclidinium bromide 10 mM solution in DMSO is a potent muscarinic acetylcholine receptor antagonist provided for laboratory research use. The ready-to-use solution supports in vitro receptor pharmacology and biochemical assays.

  • Provided as a 10 mM solution in DMSO.
  • Volume: 1 mL.
  • CAS number: 869113-09-7.
  • Molecular formula: C29H34BrNO2.
  • Molecular weight: 508.49 g·mol⁻¹.
  • Target: muscarinic acetylcholine receptors (M1-M5).
  • Reported Ki: 0.05-0.16 nM across M1-M5.
  • Intended for research use only.

Catalog No. 50-002-32730 Supplier Medchemexpress LLC Supplier No. HY1210010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Umeclidinium bromide 10 mM solution in DMSO is a potent muscarinic acetylcholine receptor antagonist provided for laboratory research use. The ready-to-use solution supports in vitro receptor pharmacology and biochemical assays.

  • Provided as a 10 mM solution in DMSO.
  • Volume: 1 mL.
  • CAS number: 869113-09-7.
  • Molecular formula: C29H34BrNO2.
  • Molecular weight: 508.49 g·mol⁻¹.
  • Target: muscarinic acetylcholine receptors (M1-M5).
  • Reported Ki: 0.05-0.16 nM across M1-M5.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.