Learn More
Medchemexpress LLC Umeclidinium-d10 bromide | 869113-09-7 | 518.55 | 1 MG
SDP
Catalog No. 5000436265
Encompass_Preferred
Umeclidinium-d10 (bromide) is the deuterium labeled Umeclidinium bromide. Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36265 Supplier Medchemexpress LLC Supplier No. HY12100S11MG
Umeclidinium-d10 (bromide) is the deuterium labeled Umeclidinium bromide. Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.