Learn More
Cayman Chemical UmeclIdInIumbromIde 100mg
SDP
Catalog No. 502984924
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist (Kis = 0.05-0.16 nM for M1-M5); inhibits acetylcholine-induced activation of recombinant M3 receptors in CHO cell membranes (IC50 = 50 = 10 µM); inhibits methacholine-induced bronchoconstriction in mice (ED50 = 0.02 UG, intranasal)
Catalog No. 50-298-4924 Supplier Cayman Chemical Supplier No. 20700100 MG
A muscarinic acetylcholine receptor antagonist (Kis = 0.05-0.16 nM for M1-M5); inhibits acetylcholine-induced activation of recombinant M3 receptors in CHO cell membranes (IC50 = 50 = 10 µM); inhibits methacholine-induced bronchoconstriction in mice (ED50 = 0.02 UG, intranasal)