Learn More
Cayman Chemical Umeclidinium (bromide), Purity: ≥98%, CAS: 869113-09-7, 10 mg
SDP
Catalog No. 502984938
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist (Kis = 0.05-0.16 nM for M1-M5); inhibits acetylcholine-induced activation of recombinant M3 receptors in CHO cell membranes (IC50 = 50 = 10 µM); inhibits methacholine-induced bronchoconstriction in mice (ED50 = 0.02 µg, intranasal)
Catalog No. 50-298-4938 Supplier Cayman Chemical Supplier No. 2070010 MG
A muscarinic acetylcholine receptor antagonist (Kis = 0.05-0.16 nM for M1-M5); inhibits acetylcholine-induced activation of recombinant M3 receptors in CHO cell membranes (IC50 = 50 = 10 µM); inhibits methacholine-induced bronchoconstriction in mice (ED50 = 0.02 µg, intranasal)