Learn More
Medchemexpress LLC Umibecestat | 1387560-01-1 | 99.8% | 5 MG
SDP
Catalog No. 5000231890
Encompass_Preferred
Umibecestat is a potent, selective BACE-1 (beta-secretase) inhibitor used for research into Alzheimer's disease. It exhibits low-nanomolar inhibition of human and mouse BACE-1 and is supplied in small research pack sizes suitable for in vitro studies.
  • Potent BACE-1 inhibitor with IC50 ≈ 11 nM (human) and 10 nM (mouse).
  • Suitable for Alzheimer's disease research and in vitro assays.
  • High solubility in DMSO (~100 mg/mL) with ultrasonic assistance.
  • Store solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month), protected from light.
Catalog No. 50-002-31890 Supplier Medchemexpress LLC Supplier No. HY1196895MG
Umibecestat is a potent, selective BACE-1 (beta-secretase) inhibitor used for research into Alzheimer's disease. It exhibits low-nanomolar inhibition of human and mouse BACE-1 and is supplied in small research pack sizes suitable for in vitro studies.
  • Potent BACE-1 inhibitor with IC50 ≈ 11 nM (human) and 10 nM (mouse).
  • Suitable for Alzheimer's disease research and in vitro assays.
  • High solubility in DMSO (~100 mg/mL) with ultrasonic assistance.
  • Store solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month), protected from light.