Learn More
Medchemexpress LLC Umibecestat (CNP520) | 1387560-01-1 | 99.8% | 10 MG
SDP
Catalog No. 5000234639
Encompass_Preferred
Umibecestat (CNP520) is a potent and selective beta-secretase (BACE-1) inhibitor used in Alzheimer's disease research. It is supplied as a research-grade compound for in vitro biochemical and cellular studies.
  • Potent BACE-1 inhibition (IC50 ~11 nM human BACE-1).
  • High purity suitable for biochemical assays (>99.8%).
  • Available as solid quantities (e.g., 10 mg) and as 10 mM solution in DMSO.
  • Molecular formula C19H15ClF7N5O2 and molecular weight 513.80 g/mol.
  • Commonly used in BACE-1 pathway and Alzheimer's disease research.
Catalog No. 50-002-34639 Supplier Medchemexpress LLC Supplier No. HY11968910MG
Umibecestat (CNP520) is a potent and selective beta-secretase (BACE-1) inhibitor used in Alzheimer's disease research. It is supplied as a research-grade compound for in vitro biochemical and cellular studies.
  • Potent BACE-1 inhibition (IC50 ~11 nM human BACE-1).
  • High purity suitable for biochemical assays (>99.8%).
  • Available as solid quantities (e.g., 10 mg) and as 10 mM solution in DMSO.
  • Molecular formula C19H15ClF7N5O2 and molecular weight 513.80 g/mol.
  • Commonly used in BACE-1 pathway and Alzheimer's disease research.