Learn More
Medchemexpress LLC Umibecestat (CNP520) | 1387560-01-1 | 99.8% | 100 MG
SDP
Catalog No. 5000233609
Encompass_Preferred
Umibecestat (CNP520) is a potent BACE-1 (beta-secretase) inhibitor used for Alzheimer's disease research. Supplied as a 100 mg research sample with supporting documentation, including a product data sheet and safety data sheet.
  • Potent BACE-1 inhibitor with reported IC50 ~11 nM (human).
  • Molecular formula C19H15ClF7N5O2; molecular weight 513.80 g/mol.
  • High purity: 99.8%.
  • Supplied as a 100 mg package for research use only.
  • Includes product data sheet and SDS in multiple languages.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-33609 Supplier Medchemexpress LLC Supplier No. HY119689100MG
Umibecestat (CNP520) is a potent BACE-1 (beta-secretase) inhibitor used for Alzheimer's disease research. Supplied as a 100 mg research sample with supporting documentation, including a product data sheet and safety data sheet.
  • Potent BACE-1 inhibitor with reported IC50 ~11 nM (human).
  • Molecular formula C19H15ClF7N5O2; molecular weight 513.80 g/mol.
  • High purity: 99.8%.
  • Supplied as a 100 mg package for research use only.
  • Includes product data sheet and SDS in multiple languages.
  • For research use only; not for human or clinical use.