Learn More
Cayman Chemical UmIfenovIrhydrochlorIde 10mg
SDP
Catalog No. 502965951
Encompass_Preferred
A broad-spectrum antiviral agent; inhibits the replication of H5N1 influenza (IC50 = 30 UG/ml) and SARS-CoV-2 in vitro (IC50 = 3.537 µM); inhibits entry of HCV pseudoparticles in Huh7 cells (IC50 = 6 UG/ml); reduces the formation of influenza-induced lung lesions in ferrets at 45 mg/kg
Catalog No. 50-296-5951 Supplier Cayman Chemical Supplier No. 1693310 MG
A broad-spectrum antiviral agent; inhibits the replication of H5N1 influenza (IC50 = 30 UG/ml) and SARS-CoV-2 in vitro (IC50 = 3.537 µM); inhibits entry of HCV pseudoparticles in Huh7 cells (IC50 = 6 UG/ml); reduces the formation of influenza-induced lung lesions in ferrets at 45 mg/kg