Learn More
Medchemexpress LLC Umifoxolaner | 2021230-37-3 | 99.97% | 643.86 | 1 MG
SDP
Catalog No. 5000352162
Encompass_Preferred
Umifoxolaner (ML-878) is a γ-aminobutyric acid (GABA) regulated chloride channels antagonist. It functions as an anti-parasitic agent.
  • Functions as a GABA regulated chloride channels antagonist
  • Acts as an anti-parasitic agent
Catalog No. 50-003-52162 Supplier Medchemexpress LLC Supplier No. HY1395871MG
Umifoxolaner (ML-878) is a γ-aminobutyric acid (GABA) regulated chloride channels antagonist. It functions as an anti-parasitic agent.
  • Functions as a GABA regulated chloride channels antagonist
  • Acts as an anti-parasitic agent