Learn More
ATCC UMNSAH/DF-1; Embryonic Fibroblast; Chicken (Gallus gallus)
Catalog No. 502385027
Encompass
UMNSAH/DF-1 is a spontaneously immortalized chicken cell line. Primary chicken embryonic fibroblasts were dissociated and grown in culture; the fibroblasts were passaged until they began to senesce; the cells were concentrated during cell senescence to maintain about 30% to about 60% culture confluence. The cells are useful as substrates for virus propagation, recombinant protein expression, and recombinant virus production. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-12203 cited in US Pat. No. 5,672,485.
Catalog No. 50-238-5027 Supplier ATCC Supplier No. CRL3586
UMNSAH/DF-1 is a spontaneously immortalized chicken cell line. Primary chicken embryonic fibroblasts were dissociated and grown in culture; the fibroblasts were passaged until they began to senesce; the cells were concentrated during cell senescence to maintain about 30% to about 60% culture confluence. The cells are useful as substrates for virus propagation, recombinant protein expression, and recombinant virus production. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-12203 cited in US Pat. No. 5,672,485.