Learn More
Aviva Systems Biology Corp UMOD Antibody - middle region (ARP41436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UMOD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UMOD. Alternate names: ADMCKD2, FJHN, HNFJ, MCKD2, THGP, THP, HNFJ1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57853