Learn More
Aviva Systems Biology Corp UMODL1 Antibody - middle region (ARP44475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UMODL1. It was validated on Western Blot. UMODL1,uromodulin-like 1,UMODL1,uromodulin-like 1,uromodulin-like 1,olfactorin,OTTHUMP00000109312,OTTHUMP00000109313The immunogen is a synthetic peptide directed towards the middle region of human UMODL1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-146