Learn More
David Kopf Instruments Model UMP3T-1 UltraMicroPump 3 with SMARTouch™ Controller
SDP
Catalog No. NC1756393
Encompass
The UltraMicroPump 3T (UMP3T) manufactured by World Precision Instruments is a versatile pump which uses micro syringes to deliver picoliter to milliliter volumes. The pump is optimum for applications that require injections of precise and small amounts of liquid. With its touchscreen controller, UMP3 can displace as little as 0.53 μL/step (using 10 μL syringe with 60 mm scale length). This item does not come with the upper bracket. The catalog number for the bracket is NC1756394 (manuf#1770C).
Catalog No. NC1756393 Supplier David Kopf Instruments Supplier No. UMP3T1
The UltraMicroPump 3T (UMP3T) manufactured by World Precision Instruments is a versatile pump which uses micro syringes to deliver picoliter to milliliter volumes. The pump is optimum for applications that require injections of precise and small amounts of liquid. With its touchscreen controller, UMP3 can displace as little as 0.53 μL/step (using 10 μL syringe with 60 mm scale length). This item does not come with the upper bracket. The catalog number for the bracket is NC1756394 (manuf#1770C).