Learn More
World Precision Instrument UMP3 UltraMicroPump -UMP3-4
SDP
Supplier:  World Precision Instrument UMP34
Encompass
With its digital controller, UltraMicroPump III can repeatably inject as little as 25nL. Syringes may be filled externally and then inserted into the pump or front-filled while mounted on the pump. 
Catalog No. 50-252-1040
Add to Cart
Request a Quote