Learn More
Aviva Systems Biology Corp UMPS Antibody - C-terminal region (ARP48147_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48147_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UMPS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UMPS. Alternate names: OPRTApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59729