Learn More
AirSea USA UN 4G box for 1 x 5L steel drum, 4G/X13/S. This 4G box is approved for shipping liquid hazardous goods.
Catalog No. NC2101159
Encompass_Preferred
UN 4G box for 1 x 5L steel drum, 4G/X13/S. This 4G box is approved for shipping liquid hazardous goods. The approved inner is AirSea Code 230.
This item is not returnable. View return policy
Catalog No. NC2101159 Supplier AirSea USA Supplier No. 730
UN 4G box for 1 x 5L steel drum, 4G/X13/S. This 4G box is approved for shipping liquid hazardous goods. The approved inner is AirSea Code 230.