Learn More
AirSea USA UN Plywood box for non-specific inners, 4DV/X107/S, Int Dims: 780mm x 380mm x 585mm
Catalog No. 501534821
Encompass_Preferred
Plywood box for non-specific inners, UN/4DV/X107/S for Hazardous or non Hazardous shipments up to 107kg/235 lbs
Catalog No. 50-153-4821 Supplier AirSea USA Supplier No. 1040
Plywood box for non-specific inners, UN/4DV/X107/S for Hazardous or non Hazardous shipments up to 107kg/235 lbs