Learn More
AirSea USA UN3373 Packaging – Bioshipper 1, suitable for Biological Substances, Category B UN3373, Packing Instruction P650. The Bioshipper-1 is capable of carrying up to 4 x BioBag-1
Catalog No. NC2101119
Encompass_Preferred
The Air Sea Containers Bioshipper-1 is supplied with 1 x 95KPa Biobag-1, 1 x Security Seal, 1 x 4 Bay Absorbent Cushioning Pouch and Packaging Instruction sheet. This product is supplied flat packed ready to ship, is designed with a fast easy pop up construction and is capable of carrying up to 4 x BioBag-1. The box dimensions allow the full internal capacity of the BioBag-1 to be utilised enhancing the economic performance of the packaging whilst reducing transport volume weight costs. The Bioshipper-1 can also be used with our Thermal Control Packaging for shipping temperature sensitive Biological Substances
This item is not returnable. View return policy
Catalog No. NC2101119 Supplier AirSea USA Supplier No. 761
The Air Sea Containers Bioshipper-1 is supplied with 1 x 95KPa Biobag-1, 1 x Security Seal, 1 x 4 Bay Absorbent Cushioning Pouch and Packaging Instruction sheet. This product is supplied flat packed ready to ship, is designed with a fast easy pop up construction and is capable of carrying up to 4 x BioBag-1. The box dimensions allow the full internal capacity of the BioBag-1 to be utilised enhancing the economic performance of the packaging whilst reducing transport volume weight costs. The Bioshipper-1 can also be used with our Thermal Control Packaging for shipping temperature sensitive Biological Substances