Learn More
AirSea USA Complete Kit for UN3373
Catalog No. 501534824
Encompass_Preferred
UN3373 Postal Pack. Our postal pack is provided as a complete kit with 95kpa Biobag, security tab seal, 4bay absorbent cushioning pouch and packaging instruction sheet. We have provided a space for the consignment details to be printed directly onto package for ease. This product is designed to be sent via the standard postal service.
Catalog No. 50-153-4824 Supplier AirSea USA Supplier No. 760
UN3373 Postal Pack. Our postal pack is provided as a complete kit with 95kpa Biobag, security tab seal, 4bay absorbent cushioning pouch and packaging instruction sheet. We have provided a space for the consignment details to be printed directly onto package for ease. This product is designed to be sent via the standard postal service.