Learn More
Centurion Scientific UN3373 MAILER-602SS DCK-12/CS
Catalog No. 50949955
Encompass_Preferred
Bioship 602Ss Dck Diagnostic Cooler Kit Has An Airtight Pressure Vessel (Secondary Container), Complete With Biohazard Label, Absorbent Pouch, Leakproof Polybag, Instructions, Cooler And Outer Fiberboard Box. This Package System Meets All Requirements Of Iata/Icao Packing Instruction 650 For Air Shipment, And All Requirements Of Domestic And International Regulations For Ground Transportation Of Diagnostic Specimens. This Kit Is For The Customer That Wants An Inexpensive Way To Ship Their Temperature Sensitive Diagnostic Specimens And Still Be In Compliance. Kits Are Sold Assembled 12 Kits/Case.
Catalog No. 50-949-955 Supplier Centurion Scientific Supplier No. 602SS DCK
Bioship 602Ss Dck Diagnostic Cooler Kit Has An Airtight Pressure Vessel (Secondary Container), Complete With Biohazard Label, Absorbent Pouch, Leakproof Polybag, Instructions, Cooler And Outer Fiberboard Box. This Package System Meets All Requirements Of Iata/Icao Packing Instruction 650 For Air Shipment, And All Requirements Of Domestic And International Regulations For Ground Transportation Of Diagnostic Specimens. This Kit Is For The Customer That Wants An Inexpensive Way To Ship Their Temperature Sensitive Diagnostic Specimens And Still Be In Compliance. Kits Are Sold Assembled 12 Kits/Case.