Learn More
Centurion Scientific UN3373 MAILER-650SS K-40/CS
Catalog No. 50949953
Encompass_Preferred
Bioship 650SS K Diagnostic Specimen Kit is a compact (6 x 4 3/8 x 3), economical, compliant for all modes of transportation, diagnostic specimen shipper. Contains an airtight pressure vessel (secondary container), biohazard label, absorbent pouch, instructions, and outer fiberboard box. This package system meets postal Domestic Mail Manual requirements and DOT 49CFR requirements for ground shipment, along with ICAO/IATA Packing Instruction 650 for air shipment of diagnostic specimens. This kit is for the customer that is looking for an easy-to-use, reusable, low-cost diagnostic specimen shipper that meets every regulation for all modes of transportation. Features: Meets the requirements of IATA/ICAO Packing Instructions 650 for shipping Diagnostic Specimens by air, as well as CFR 49 173.199 for ground transportation of these items. Secondary container has an easy open positive lock closure. Secondary container passes 95kPa test and offers high impact resistance.
Catalog No. 50-949-953 Supplier Centurion Scientific Supplier No. 650SS K
Bioship 650SS K Diagnostic Specimen Kit is a compact (6 x 4 3/8 x 3), economical, compliant for all modes of transportation, diagnostic specimen shipper. Contains an airtight pressure vessel (secondary container), biohazard label, absorbent pouch, instructions, and outer fiberboard box. This package system meets postal Domestic Mail Manual requirements and DOT 49CFR requirements for ground shipment, along with ICAO/IATA Packing Instruction 650 for air shipment of diagnostic specimens. This kit is for the customer that is looking for an easy-to-use, reusable, low-cost diagnostic specimen shipper that meets every regulation for all modes of transportation. Features: Meets the requirements of IATA/ICAO Packing Instructions 650 for shipping Diagnostic Specimens by air, as well as CFR 49 173.199 for ground transportation of these items. Secondary container has an easy open positive lock closure. Secondary container passes 95kPa test and offers high impact resistance.