Learn More
Unchained Labs UNA, 100 KDA MWCO, CASE OF 40
SDP
Supplier:  Unchained Labs 9012003
Encompass
Una consumable for Unagi instrument, 0.5 - 8 mL capacity.MWCO: 100kDa, regenerated cellulose (RC). 8 Unas per box,5 boxes, case total of 40.
Catalog No. NC2210873