Learn More
Unchained Labs UNA, 30 KDA MWCO, CASE OF 200
SDP
Catalog No. NC2210895
Encompass
Una consumable for Unagi instrument, 0.5 - 8 mL capacity.MWCO: 30kDa, regenerated cellulose (RC). 8 Unas per box,25 boxes, case total of 200
This item is not returnable. View return policy
Catalog No. NC2210895 Supplier Unchained Labs Supplier No. 9012012
Add to Cart
Add to Cart
Request a Quote
Una consumable for Unagi instrument, 0.5 - 8 mL capacity.MWCO: 30kDa, regenerated cellulose (RC). 8 Unas per box,25 boxes, case total of 200