Learn More
Medchemexpress LLC UNC 0631 (Standard) | 1320288-19-4 | 99.5% | 635.93 | 1 MG
SDP
Catalog No. 5000347771
Encompass_Preferred
UNC 0631 (Standard) is an analytical standard and a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. It reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM. This product is intended for research and analytical applications.
  • Analytical standard for UNC 0631
  • Potent histone methyltransferase G9a inhibitor
  • Reduces H3K9me2 levels in MDA-MB-231 cells
  • Intended for research and analytical applications
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-003-47771 Supplier Medchemexpress LLC Supplier No. HY13808R1MG
UNC 0631 (Standard) is an analytical standard and a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. It reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM. This product is intended for research and analytical applications.
  • Analytical standard for UNC 0631
  • Potent histone methyltransferase G9a inhibitor
  • Reduces H3K9me2 levels in MDA-MB-231 cells
  • Intended for research and analytical applications
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS