Learn More
Eurofins Discoverx UNC 0638, 10 mg. (92-1150)
Supplier:  Eurofins Discoverx 921150
Encompass
UNC 0638 is a cell permeable synthetic small molecule inhibitor of the G9a and GLP histone lysine methyltransferase enzymes (IC50 of <15 nM and 19 nM, respectively), which contribute to early embryogenesis, genomic imprinting, and lymphocyte development. >1000-fold selective vs other HMTases. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9681