Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UNC-2170 100mg | 1648707-58-7 | 313.23 | C14H21BrN2O | 100 MG
SDP

Catalog No. 5000226301
Encompass_Preferred

UNC-2170 is a fragment-like small-molecule ligand of 53BP1 used as a biochemical probe in DNA damage repair research. It binds selectively to 53BP1 and is applied to study recruitment to double-strand break sites.

  • CAS number 1648707-58-7.
  • Molecular formula C14H21BrN2O.
  • Molecular weight 313.23.
  • Purity 98.84% (as supplied).
  • Appearance viscous liquid, colorless to light yellow.
  • Target 53BP1; pathway DNA damage repair.
  • Storage solid at 4°C, protect from light; in solvent store at -80°C for long term.

Catalog No. 50-002-26301 Supplier Medchemexpress LLC Supplier No. HY115531100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UNC-2170 is a fragment-like small-molecule ligand of 53BP1 used as a biochemical probe in DNA damage repair research. It binds selectively to 53BP1 and is applied to study recruitment to double-strand break sites.

  • CAS number 1648707-58-7.
  • Molecular formula C14H21BrN2O.
  • Molecular weight 313.23.
  • Purity 98.84% (as supplied).
  • Appearance viscous liquid, colorless to light yellow.
  • Target 53BP1; pathway DNA damage repair.
  • Storage solid at 4°C, protect from light; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.