Learn More
Medchemexpress LLC UNC-2170 10mg | 1648707-58-7 | 313.23 g/mol | C14H21BrN2O | 10 MG
SDP
Catalog No. 5000307962
Encompass_Preferred
UNC-2170 is a fragment-like small-molecule ligand that binds the methyl-lysine (Kme) recognition domain of 53BP1 and is used as a research tool to study DNA damage repair pathways. It shows moderate affinity for 53BP1 (IC50 = 29 μM; Kd = 22 μM) and selectivity versus other Kme reader proteins.
  • Fragment-like ligand for 53BP1 with selectivity versus other Kme readers.
  • Moderate affinity: IC50 = 29 μM; Kd = 22 μM.
  • Molecular formula C14H21BrN2O; molecular weight 313.23 g/mol.
  • High purity (≈98.8%) and viscous liquid appearance.
  • Supplied as a 10 mg research reagent; store protected from light.
Catalog No. 50-003-07962 Supplier Medchemexpress LLC Supplier No. HY11553110MG
UNC-2170 is a fragment-like small-molecule ligand that binds the methyl-lysine (Kme) recognition domain of 53BP1 and is used as a research tool to study DNA damage repair pathways. It shows moderate affinity for 53BP1 (IC50 = 29 μM; Kd = 22 μM) and selectivity versus other Kme reader proteins.
  • Fragment-like ligand for 53BP1 with selectivity versus other Kme readers.
  • Moderate affinity: IC50 = 29 μM; Kd = 22 μM.
  • Molecular formula C14H21BrN2O; molecular weight 313.23 g/mol.
  • High purity (≈98.8%) and viscous liquid appearance.
  • Supplied as a 10 mg research reagent; store protected from light.