Learn More
Active Motif UNC-2170
SDP
Catalog No. NC1841443
Encompass
Active Motif's The DNA damage response protein 53BP1 utilizes its tandem tudor domain to recognize dimethylated lysine 20 on histone (H4K20me2), a modification associated with double-stranded DNA breaks. UNC-2170, identified by screening, was found to be a uM ligand for 53BP1 which also demonstrated at least 17-fold selectivity for 53BP1 over other methyl-lysine binding proteins. The compound functions as 53BP1 antagonist in cell lysates and suppresses class switch recombination in whole cells, a process requiring a functioning 53BP1 tudor domain.
Catalog No. NC1841443 Supplier Active Motif Supplier No. 14120
Active Motif's The DNA damage response protein 53BP1 utilizes its tandem tudor domain to recognize dimethylated lysine 20 on histone (H4K20me2), a modification associated with double-stranded DNA breaks. UNC-2170, identified by screening, was found to be a uM ligand for 53BP1 which also demonstrated at least 17-fold selectivity for 53BP1 over other methyl-lysine binding proteins. The compound functions as 53BP1 antagonist in cell lysates and suppresses class switch recombination in whole cells, a process requiring a functioning 53BP1 tudor domain.