Learn More
TARGETMOL CHEMICALS INC UNC-2170 5MG
SDP
Catalog No. 502580778
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UNC-2170 (UNC2170 Maleate) is the methyl-lysine binding protein 53BP1 ligand. purity: 98%
Catalog No. 50-258-0778 Supplier TARGETMOL CHEMICALS INC Supplier No. T249255MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UNC-2170 (UNC2170 Maleate) is the methyl-lysine binding protein 53BP1 ligand. purity: 98%