Learn More
Medchemexpress LLC UNC-2170 5mg | 1648707-58-7 | 5 MG
SDP
Catalog No. 5000228051
Encompass_Preferred
This small-molecule ligand selectively binds the methyl-lysine reader protein 53BP1 and is used as a research tool in DNA damage repair studies. It exhibits micromolar binding and inhibitory activity and is supplied as a high-purity solid for laboratory use.
  • Selective 53BP1 ligand with micromolar potency (IC50 29 μM; Kd 22 μM).
  • Demonstrates ≥17-fold selectivity versus other methyl-lysine reader proteins.
  • High reported purity of 98.84% suitable for screening.
  • Available in small solid quantities for biochemical assays.
  • Recommended storage: solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-28051 Supplier Medchemexpress LLC Supplier No. HY1155315MG
This small-molecule ligand selectively binds the methyl-lysine reader protein 53BP1 and is used as a research tool in DNA damage repair studies. It exhibits micromolar binding and inhibitory activity and is supplied as a high-purity solid for laboratory use.
  • Selective 53BP1 ligand with micromolar potency (IC50 29 μM; Kd 22 μM).
  • Demonstrates ≥17-fold selectivity versus other methyl-lysine reader proteins.
  • High reported purity of 98.84% suitable for screening.
  • Available in small solid quantities for biochemical assays.
  • Recommended storage: solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).