Learn More
Selleck Chemical LLC UNC-926 S5262-10mg
SDP
Catalog No. 503112006
Encompass_Preferred
UNC-926 is a L3MBTL1 domain inhibitor with Kd value of 3 9 M for binding with the MBT domain of the L3MBTL1 protein
Catalog No. 50-311-2006 Supplier Selleck Chemical LLC Supplier No. S526210MG
UNC-926 is a L3MBTL1 domain inhibitor with Kd value of 3 9 M for binding with the MBT domain of the L3MBTL1 protein