Learn More
Cayman Chemical UNC-CA2-103 5mg
SDP
Catalog No. 502944719
Encompass_Preferred
A TLK2 inhibitor (IC50 = 18 nM); selective for TLK2 over TLK1, TrkC, and FLT3 at 500 nM and a panel of 468 kinases at 1 µM, but it does inhibit 11 kinases by >90% in the same panel
Catalog No. 50-294-4719 Supplier Cayman Chemical Supplier No. 412415 MG
A TLK2 inhibitor (IC50 = 18 nM); selective for TLK2 over TLK1, TrkC, and FLT3 at 500 nM and a panel of 468 kinases at 1 µM, but it does inhibit 11 kinases by >90% in the same panel