Learn More
Selleck Chemical LLC UNC0379 5mg
SDP
Supplier:  Selleck Chemical LLC S75705MG
Encompass_Preferred
UNC0379 is a selective, substrate competitive inhibitor of N-lysine methyltransferase SETD8 with IC50 of 7.9 ?M, high selectivity over 15 other methyltransferases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3287