Learn More
Medchemexpress LLC UNC0379 trifluoroacetate | 1620401-83-3 | MFCD28167817 | 99.9% | 527.58 g·mol⁻¹ | C25H36F3N5O4 | 5MG
SDP
Catalog No. 5000239055
Encompass_Preferred
UNC0379 trifluoroacetate is the trifluoroacetate salt of UNC0379, a selective substrate-competitive inhibitor of the lysine methyltransferase SETD8 (KMT5A). It is supplied as a high-purity research reagent for biochemical and cellular studies, with reported activity IC50 = 7.3 μM and KD = 18.3 μM. For laboratory research use only.
Catalog No. 50-002-39055 Supplier Medchemexpress LLC Supplier No. HY12335A5MG
UNC0379 trifluoroacetate is the trifluoroacetate salt of UNC0379, a selective substrate-competitive inhibitor of the lysine methyltransferase SETD8 (KMT5A). It is supplied as a high-purity research reagent for biochemical and cellular studies, with reported activity IC50 = 7.3 μM and KD = 18.3 μM. For laboratory research use only.