Learn More
Cayman Chemical UNC0631, 10 mg
SDP
Supplier:  Cayman Chemical 1108410
Encompass_Preferred
UNC0631 is a potent and selective inhibitor of G9a activity in vitro (IC50 = 4 nM) and G9a/GLP-mediated dimethylation of histone 3 on lysine 9 in MDA-MB-231 cells (IC50 = 25 nM).1 It potently inhibits G9a/GLP activity in a variety of cancer cell lines as well as in the human fetal lung IMR-90 cell line.1WARNING This product is not for human or veterinary use.
Catalog No. NC2819200