Learn More
Apexbio Technology LLC UNC0638, 10mg. Cas: 1255580-76-7 MFCD: MFCD22208587. G9a/GLP HMTase inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC A191410
Encompass_Preferred
UNC0638 is a potent and selective inhibitor of G9a and GLP with IC50 values of < 15 nM and 19 1 nM, respectively [1].GLP forms a heterodimer with G9a. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4723