Learn More
Medchemexpress LLC UNC10217938A | 1347749-97-6 | 99.75% | 456.54 | 100 MG
SDP
Catalog No. 5000348775
Encompass_Preferred
UNC10217938A is a 3-deazapteridine analog with strong oligonucleotide enhancing effects. It enhances oligonucleotide effects by modulating their intracellular trafficking and release from endosomes, and also enhances the effects of antisense and siRNA oligonucleotides.
  • Enhances luciferase induction in HelaLuc705 cells
  • Enhances effects of receptor-targeted oligonucleotide conjugates, uncharged morpholino oligonucleotides, and negatively charged oligonucleotides
  • Causes partial release of oligonucleotide from late endosomes to the cytosol followed by nuclear accumulation
  • Systemic treatment produces distinct increases in EGFP fluorescence in liver, kidney, and heart in EGFP654 mice
Catalog No. 50-003-48775 Supplier Medchemexpress LLC Supplier No. HY136151100MG
UNC10217938A is a 3-deazapteridine analog with strong oligonucleotide enhancing effects. It enhances oligonucleotide effects by modulating their intracellular trafficking and release from endosomes, and also enhances the effects of antisense and siRNA oligonucleotides.
  • Enhances luciferase induction in HelaLuc705 cells
  • Enhances effects of receptor-targeted oligonucleotide conjugates, uncharged morpholino oligonucleotides, and negatively charged oligonucleotides
  • Causes partial release of oligonucleotide from late endosomes to the cytosol followed by nuclear accumulation
  • Systemic treatment produces distinct increases in EGFP fluorescence in liver, kidney, and heart in EGFP654 mice