Learn More
Medchemexpress LLC HY-136151 5mg Medchemexpress, UNC10217938A CAS:1347749-97-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1361515MG
Encompass_Preferred
Medchemexpress, HY-136151 5mg UNC10217938A CAS:1347749-97-6 UNC10217938A is a 3-deazapteridine analog with strong oligonucleotide enhancing effects. UNC10217938A enhances oligonucleotides effects by modulating their intracellular trafficking and release from endosomes. UNC10217938A also enhances the effects of antisense and siRNA oligonucleotides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0400