Learn More
TARGETMOL CHEMICALS INC UNC1999 25MG
SDP
Catalog No. 502580406
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UNC1999 is a orally bioavailable effective and specific inhibitor of EZH2 (IC50=2 nM) and EZH1 (IC50=45). purity: 99%
Catalog No. 50-258-0406 Supplier TARGETMOL CHEMICALS INC Supplier No. T305725MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UNC1999 is a orally bioavailable effective and specific inhibitor of EZH2 (IC50=2 nM) and EZH1 (IC50=45). purity: 99%