Learn More
Selleck Chemical LLC UNC1999 5mg 1431612-23-5
SDP
Supplier:  Selleck Chemical LLC S71655MG
Encompass_Preferred
UNC1999 is a potent, orally bioavailable and selective inhibitor of EZH2 and EZH1 with IC50 of 2 nM and 45 nM in cell-free assays, respectively, showing >1000-fold selectivity over a broad range of epigenetic and non-epigenetic targets. UNC1999 is a potent autophagy inducer. UNC1999 specifically suppresses H3K27me3/2 and induces a range of anti-leukemia effects including anti-proliferation, differentiation, and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1267212