Learn More
Selleck Chemical LLC UNC2025 HCl 25mg 2070015-17-5 UNC2025 hydrochloride
SDP
Catalog No. 501364969
Encompass_Preferred
UNC2025 HCl is a potent and orally bioavailable dual MER/FLT3 inhibitor with IC50 of 0.74 nM and 0.8 nM, respectively, about 20-fold selectivity over Axl and Tyro3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4969 Supplier Selleck Chemical LLC Supplier No. S757625MG
UNC2025 HCl is a potent and orally bioavailable dual MER/FLT3 inhibitor with IC50 of 0.74 nM and 0.8 nM, respectively, about 20-fold selectivity over Axl and Tyro3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.